Learn More
Medchemexpress LLC Pralnacasan | 192755-52-5 | MFCD18381953 | 99.8% | 523.54 g·mol⁻1 | C26H29N5O7 | 10 MG
SDP
Supplier:  Medchemexpress LLC HY1967610MG
Encompass_Preferred
Pralnacasan is a potent, selective, orally active inhibitor of interleukin-1β converting enzyme (caspase-1) used in preclinical inflammation and apoptosis research. It is supplied as a high-purity white to off-white powder with defined solubility and storage recommendations for in vitro and in vivo studies.
  • High potency with Ki of 1.4 nM against caspase-1.
  • High purity (99.8%) suitable for research applications.
  • Supplied as a white to off-white solid powder for easy handling.
  • Highly soluble in DMSO (220 mg/mL) for stock preparation.
  • Formulation protocols available for in vivo dosing to achieve clear solutions.
  • Stable when stored as powder at -20°C for extended periods.
Catalog No. 50-002-98686