Learn More
Medchemexpress LLC Pralsetinib | 2097132-94-8 | 99.98% | 533.60 | 100 MG
SDP
Supplier:  Medchemexpress LLC 100MGHY112301
Encompass_Preferred
Pralsetinib (BLU-667) is a potent, selective RET kinase inhibitor. It targets wild-type RET and various mutants (V804L, V804M, M918T) and the CCDC6-RET fusion, with IC50s from 0.3 to 0.4 nM. For research use only, it inhibits NSCLC and thyroid cancer xenograft growth driven by RET mutations and fusions, without inhibiting VEGFR-2. Dose-dependent activity is seen in KIF5B-RET Ba/F3 and V804L Ba/F3 allograft tumors.
  • Potent and selective RET inhibitor.
  • Targets wild-type RET and specific mutants/fusions.
  • Inhibits NSCLC and thyroid cancer xenograft growth.
  • For research use only.
Catalog No. 50-003-64757