Learn More
Selleck Chemical LLC Praluzatamab ravtansine-1MG-5
SDP
Supplier:  Selleck Chemical LLC D40391MG5
Encompass_Preferred
Praluzatamab ravtansine (CX-2009) is a conditionally activated Probody drug conjugate (PDC) comprising an anti-CD166 mAb conjugated to DM4 with a protease-cleavable linker and a peptide mask that limits target engagement in normal tissue and circulation
Catalog No. 50-313-6023