Learn More
Medchemexpress LLC HY-B0410 500mg Medchemexpress, Pramipexole CAS:104632-26-0 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYB0410500MG
Encompass_Preferred
Medchemexpress, HY-B0410 500mg Pramipexole CAS:104632-26-0 Pramipexole is a selective and blood-brain barrier (BBB) penetrant dopamine D2-type receptor agonist, with Kis of 2.2 nM, 3.9 nM, 0.5 nM and 1.3 nM for D2-type receptor, D2, D3 and D4 receptors, respectively. Pramipexole can be used for the research of Parkinson's disease (PD) and restless legs syndrome (RLS). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2524