Learn More
Medchemexpress LLC Pramipexole dihydrochloride hydrate | 191217-81-9 | 98.0% | 1 ML
SDP
Supplier:  Medchemexpress LLC HYB0410A10MM/1ML
Encompass_Preferred
Pramipexole dihydrochloride hydrate is a selective and blood-brain barrier (BBB) penetrant dopamine D2-type receptor agonist, with Kis of 2.2 nM, 3.9 nM, 0.5 nM, and 1.3 nM for D2-type receptor, D2, D3, and D4 receptors, respectively. It can be used for the research of Parkinson's disease (PD) and restless legs syndrome (RLS).
  • Selective and blood-brain barrier penetrant dopamine D2-type receptor agonist
  • Used for research of Parkinson's disease (PD) and restless legs syndrome (RLS)
  • Shows low binding affinity for D1-type receptor (>50,000 nM IC50)
  • Attenuates levodopa-induced toxicity in mesencephalic cultures
  • Significantly reduces infarction volume and improves neurological recovery
Catalog No. 50-004-06782