Learn More
Apexbio Technology LLC Pramoxine HCl 637-58-1 10mM (in 1mL DMSO)
SDP
Supplier:  Apexbio Technology LLC B18195.1
Encompass_Preferred
Pramoxine HCl is a local anesthetic that transiently inhibits sensory nerve signal conduction through sodium channel blockade in neuronal membranes thereby reducing local sensory nerve impulses and producing anesthetic and antipruritic effects Pramoxine HCl exerts its biological activity primarily through sodium channel blockade Based on these pharmacological properties Pramoxine HCl holds research potential in the investigation of sensory nerve inhibition mechanisms pruritus management and in vitro nerve conduction assays
Catalog No. 50-000-92685