Learn More
Medchemexpress LLC Pranidipine (OPC-13340) | 99522-79-9 | 99.5% | 448.47 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY19664100MG
Encompass_Preferred
Pranidipine (OPC-13340) is a potent, long-acting 1,4-dihydropyridine calcium channel blocker with antihypertensive activity. It is for research use only.
  • Potent, long-acting 1,4-dihydropyridine calcium channel blocker
  • Exhibits antihypertensive activity
  • For research use only
  • Shows antagonist activity at rabbit Cav1.2 in HEK293 cells with an IC50 of 37 nM
  • Shows antagonist activity at rat Cav1.3 in HEK293 cells with an IC50 of 37 nM
Catalog No. 50-004-02719