Learn More
Medchemexpress LLC Pranlukast (hemihydrate) | 150821-03-7 | 99.3% | 1 ML
SDP
Supplier:  Medchemexpress LLC HYB0290A10MM/1ML
Encompass_Preferred
Pranlukast hemihydrate is a highly potent, selective, and competitive antagonist of peptide leukotrienes. It is suitable for laboratory research and inhibits [3H]LTE4, [3H]LTD4, and [3H]LTC4 bindings to lung membranes with Ki values of 0.63±0.11, 0.99±0.19, and 5640±680 nM, respectively.
  • White to off-white solid appearance
  • Stable under recommended storage conditions
  • For research use only
Catalog No. 50-004-05874