Learn More
Selleck Chemical LLC Prasugrel 25mg 150322-43-3 CS-747, LY640315
SDP
Supplier:  Selleck Chemical LLC S125825MG
Encompass_Preferred
Prasugrel (CS-747, LY640315) is a thienopyridine ADP receptor (P2Y12) antagonist, used for the reduction of thrombotic cardiovascular events. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-783-9