Learn More
Medchemexpress LLC Prasugrel hydrochloride | 389574-19-0 | C20H21ClFNO3S | 500 MG
SDP
Supplier:  Medchemexpress LLC HY15284A500MG
Encompass_Preferred
Prasugrel hydrochloride (PCR 4099 hydrochloride) is a thienopyridine and a proagent that inhibits platelet function. It is an orally active and potent P2Y12 receptor antagonist, which works by inhibiting ADP-induced platelet aggregation. In rat platelets, the active metabolite of Prasugrel hydrochloride inhibits in vitro platelet aggregation induced by adenosine ADP (10μM) with an IC50 value of 1.8 μM.
  • Exhibits faster and significantly more potent antiplatelet effects in vivo compared to Clopidogrel.
  • Requires in vivo metabolic processing as an inactive prodrug to generate its active antiplatelet metabolite.
  • Rapidly absorbed from the gut following oral administration.
  • Achieves maximum plasma levels of active metabolite within 1 hour.
  • Leads to effective, maximum inhibition of platelet aggregation within 1-2 hours.
Catalog No. 50-003-73800