Learn More
Apexbio Technology LLC Pravastatin 81093-37-0 10mg
SDP
Supplier:  Apexbio Technology LLC B112510
Encompass_Preferred
Pravastatin (CAS 81093-37-0) is a small molecule inhibitor of 3-hydroxy-3-methyl-glutaryl-coenzyme A (HMG-CoA) reductase the rate-limiting enzyme in the mevalonate pathway of cholesterol biosynthesis It exhibits an inhibitory concentration (IC50) of 5 6 M against sterol synthesis Pravastatin is widely utilized in biomedical research to investigate cholesterol metabolism the regulation of lipid pathways and the physiological and pathological roles of HMG-CoA reductase inhibition Its application supports studies on cardiovascular disease models and the broader implications of statin-mediated modulation of cellular cholesterol homeostasis
Catalog No. 50-000-94160