Learn More
Selleck Chemical LLC Pravastatin sodium-50mg
SDP
Supplier:  Selleck Chemical LLC S303650MG
Encompass_Preferred
Pravastatin sodium (CS-514) is an HMG-CoA reductase inhibitor against sterol synthesis with IC50 of 5.6 μM.
Catalog No. 50-238-9968