Learn More
Medchemexpress LLC Pravastatin sodium | 81131-70-6 | 99.4% | 200 MG
SDP
Supplier:  Medchemexpress LLC HYB0165A200MG
Encompass_Preferred
Pravastatin sodium is an HMG-CoA reductase inhibitor that acts against sterol synthesis, with an IC50 of 5.6 μM. It belongs to the statin drug class and is utilized alongside diet, exercise, and weight-loss strategies to lower cholesterol and help prevent cardiovascular disease.
  • Functions as an HMG-CoA reductase inhibitor.
  • Targets sterol synthesis.
  • Exhibits an IC50 of 5.6 μM for HMG-CoA reductase.
  • Demonstrates inhibitory activity against cellular steroidogenesis in HepG2 cells with an IC50 of 1.37 μM.
  • Inhibits cholesterol synthesis from [14C]-Acetate in isolated rat hepatocytes with an IC50 of 0.1 μM.
  • Aids in lowering cholesterol.
  • Contributes to the prevention of cardiovascular disease.
Catalog No. 50-004-06463