Learn More
Medchemexpress LLC Pravastatin sodium | 81131-70-6 | 99.4% | 5 MG
SDP
Supplier:  Medchemexpress LLC HYB0165A5MG
Encompass_Preferred
Pravastatin sodium is an HMG-CoA reductase inhibitor that acts against sterol synthesis. It belongs to the statin drug class and is used to lower cholesterol and prevent cardiovascular disease in conjunction with diet, exercise, and weight-loss.
  • HMG-CoA reductase inhibitor
  • IC50 of 5.6 μM against sterol synthesis
  • Lowers cholesterol
  • Prevents cardiovascular disease
  • Inhibits cellular steroidgenesis in HepG2 cells with an IC50 of 1.37 μM
  • Inhibits cholesterol synthesis in rat hepatocytes with an IC50 of 0.1 μM
Catalog No. 50-004-06316