Learn More
Medchemexpress LLC Pravastatin sodium (CS-514 sodium) | 81131-70-6 | 99.38% | 10 G
SDP
Supplier:  Medchemexpress LLC HYB0165A10G
Encompass_Preferred
Pravastatin sodium (CS-514 sodium) is an HMG-CoA reductase inhibitor, primarily used for research purposes. It targets sterol synthesis with an IC50 of 5.6 μM and belongs to the statin drug class. It is indicated for lowering cholesterol and preventing cardiovascular disease when used in combination with diet, exercise, and weight-loss.
  • HMG-CoA reductase inhibitor
  • Targets sterol synthesis with an IC50 of 5.6 μM
  • Used for research purposes
  • Inhibits cellular steroidogenesis in HepG2 cells with an IC50 of 1.37 μM
  • Inhibits cholesterol synthesis from [14C]-Acetate in rat hepatocytes with an IC50 of 100 nM
Catalog No. 50-004-07247