Learn More
Medchemexpress LLC Praziquantel-d11 | 1246343-36-1 | 99.6% | 500 UG
SDP
Supplier:  Medchemexpress LLC HYB0244S500UG
Encompass_Preferred
Praziquantel-d11 is the deuterium labeled Praziquantel, which is an anthelmintic. This compound is intended for research use only.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
Catalog No. 50-004-05865