Learn More
Cayman Chemical PrazIquntel 10g
SDP
Supplier:  Cayman Chemical 2050810 G
Encompass_Preferred
An anthelmintic agent; lethal to S. mansoni schistosomula and adults (LC50s = 0.68 and 0.03 mg/ml, respectively); reduces worm burden in a mouse model of S. mansoni infection at 172 and 592 mg/kg
Catalog No. 50-298-0905