Learn More
Malvern Panalytical PRE FUSED FLUX LITH TETR/L 1KG
Supplier:  Malvern Panalytical C060170
Encompass
Ultra high purity 99.995+ pre fused flux to prepare samples for XRF or ICP. Lithium Tetraborate/Lithium Iodide 99.50/0.50. High Density, Grade Ultra Pure. 1kg.
Catalog No. NC2468729