Learn More
Analytical Sales & Services Analytical Sales & Services Pre-Slit Silicone/PTFE Cap Mat, Excellent Chemical Resistance, Pre-Slit Design, 5 Units
Supplier: Analytical Sales & Services 35085
The Pre-Slit Silicone/PTFE Cap Mat offers precise sealing for scientific applications, ensuring sample integrity and minimizing contamination. Its dual-layer design merges the chemical inertness of PTFE with silicone's resealing properties, ideal for high-throughput storage. The pre-slit feature allows easy needles access, distinguishing it from other products. Perfectly compatible with 350µL Select-A-Vial™ Rack Systems, it's designed for efficient sample handling in laboratory environments.