Learn More
Chemglass Life Sciences Preassembled Closure and Septa, Blue, 10-425, 0.040" PTFE/Silicone Septa
SDP
Supplier:  Chemglass Life Sciences CV5190B010
Encompass_Preferred
Preassembled Closure and Septa, Blue, 10-425, 0.040" PTFE/Silicone Septa. Pre-assembled closure and septa minimize contamination from handling. They are convenient and available in a variety of colors with your choice of septa.
Catalog No. 50-231-261