Learn More
Perkin Elmer US LLC Pre-Assembled Magnetic Aluminum Screw Cap with Ultra-Low Bleed PTFE/Silicone (Blue/White) Septa - 1.5 mm thick, 100/pk
Supplier:  Perkin Elmer US LLC N9304175
Encompass_Preferred
Cap Type Cap Material Color Septa Material Septa Thickness Septa Features Quantity Magnetic Screw Top Aluminum Blue/White PTFE/Silicone 1.5 mm Ultra-Low Bleed 100
Catalog No. 50-211-2624