Learn More
Cayman Chemical Precellys 2 mL Soft Tissue H
SDP
Supplier:  Cayman Chemical 1001115210 BOX
Encompass_Preferred
Kit Includes 50 preps prefilled with 1.4 mm ceramic (zirconium oxide) beads in 2 mL standard tubes. CK14 is designed for soft tissue homogenization like brain, liver, kidney, skin, plant leaves, mammalian cells, and more. Sample size range from 20 mg to 200 mg tissue or 200 µL to 1.6 mL culture. [Bertin Catalog P000912-LYSK0-A.0; Previously KT03961-1-003.2]
Catalog No. 50-288-6846