Learn More
Cayman Chemical Precellys 7 mL Soft Tissue H
SDP
Supplier:  Cayman Chemical 152385 BOX
Encompass_Preferred
Kit includes 50 preps of 1.4 mm ceramic (zirconium oxide) beads in 7 mL standard tubes with O-ring. CK14 7 mL is designed for soft tissue homogenization like rat cerebelum, skin and more. Sample size range from 200 mg to 2 g tissue or 200 µL to 6 mL culture. [Bertin Catalog P000940-LYSK0-A.0; Previously KT03961-1-307.7]
Catalog No. 50-297-2788