Learn More
Bertin Corp Bacteria Lysing CK01 - 7mL tubes 50 prep
SDP
Supplier:  Bertin Corp P000938LYSK0A.0
Encompass_Preferred
Lysing kit for Precellys - CK01 Micro-organism kit - 7mL tubes 50 prep. The CK01 matrix is composed of 0.1mm ceramic (zirconium oxide) beads and is designed for micro-organisms lysing like gram- and gram+ bacteria but also spores, micro-algae etc. It ensures optimal extraction yields of DNA/RNA/Protein for samples from 20mg to 200mg of tissue. The kits requires the use of the dedicated tube holder for Precellys Evolution (SP02520-810-NC010.0)
Catalog No. 50-154-2956