Learn More
Cayman Chemical Precellys Zirconium Oxide Be
SDP
Supplier:  Cayman Chemical 104021 BOX
Encompass_Preferred
Includes 500 preps of 1.4 mm ceramic beads. These beads are designed for use with Precellys Tissue Homogenizers. Beads are designed for soft tissue homogenization like brain, liver, kidney, and more. [Bertin Catalog P000927-LYSK0-A.0; Previously KT03961-1-103.BK]
Catalog No. 50-288-6839