Learn More
Techni Tool Inc PRECISION ERGONOMIC TWEEZERS T
SDP
Supplier:  Techni Tool Inc TL 7ASAET
Encompass
Product DetailsType 7ATip description: fine, curvedBody material: anti-magnetic, anti-acid stainless steelHandle coating: ESD thermoplastic rubberProfessional Lindström precision tweezers offer perfect balance, tip alignment and symmetryMade in Switzerland
Catalog No. NC3953266