Learn More
Peak Scientific Instruments Ltd PRECISION HYDROGEN 100CC
SDP
Supplier:  Peak Scientific Instruments Ltd 630100
Encompass_Preferred
  • Robust and reliable hydrogen gas generator designed for laboratory applications.
  • Safer and more convenient alternative to traditional gas cylinders.
  • Ideal for supplying hydrogen fuel gas to Gas Chromatography detectors.
  • Utilizes Proton Exchange Membrane (PEM) technology for on-demand hydrogen production.
  • Incorporates a desiccant filtration stage for dry gas delivery.
  • Features internal leak detection with automatic shutdown for enhanced safety.
  • Minimal maintenance requirements including de-ionizer cartridge replacement.
  • Compact and stackable design optimizes lab bench space.
  • Produces high-purity hydrogen with a maximum purity of 99.9995%.
  • Max flow rate of 100 cc/min and an output pressure of 100 psi.
  • Operates at an ambient temperature of 10°C - 35°C.
  • Electrical requirement of 100-230 V, 50/60 Hz.
Catalog No. NC1882020
Only null left
Add to Cart
Request a Quote