Learn More
MOUSER ELECTRONICS Metal Film Resistors, 250 ohms
Supplier:  MOUSER ELECTRONICS MBB0207VD2500BC1
Encompass
0.1% Tolerance, 400 mW Power Rating, 25 PPM /C Temp Coefficient, and -55/+125 C Min/Max Operating Temp.
Catalog No. NC1766986