Learn More
World Precision Instrument Precision Stereo Zoom Binocular Microscope (IV) on Track Stand
SDP
Supplier:  World Precision Instrument PZMIV
Encompass
The fourth generation of WPI’s precision stereo zoom microscopes uses modular, two parallel beam path design and high quality optical system. The advanced optical design with planachromatic objectives provides sharp and distortion-free contrast image throughout the entire zoom range and comes with an impressive 5-year warranty. The microscope offers planachromatic objectives the ensure no optical distortion. It has a large zoom ratio of 8:1, and a large zoom range of 0.62x-5.0x. It has step and continuous zoom and comes with a 5-year warranty.
Catalog No. 50-253-8614