Learn More
Bound Tree Medical PRECISION XTRA GLUCOMETER
Supplier:  Bound Tree Medical 179837
Encompass_Preferred
The Precision Xtra features easy two-step testing; simply insert the test strip and add a drop of blood. Special end-fill / top-fill test strip design and alternative-site capability make glucose testing more convenient and flexible. The Precision Xtra now offers a faster test time and requires a 40 percent smaller sample size. Stores up to 450 test results with date and time in its memory. Permits second drop of blood if first sample is not adequate. Test only starts with sufficient sample, minimizing error messages and repeat test. Reads glucose levels from 20 - 500mg/dL. The Precision Xtra features easy two-step testing; simply insert the test strip and add a drop of blood. Special end-fill / top-fill test strip design and alternative-site capability make glucose testing more convenient and flexible. The Precision Xtra now offers a faster test time and requires a 40 percent smaller sample size. Stores up to 450 test results with date and time in its memory. Permits second drop of
Catalog No. NC0288721
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.