Learn More
Cayman Chemical PrecoatedMouse ntI-RabbIt 5ea
SDP
Supplier:  Cayman Chemical 4000065 EA
Encompass_Preferred
A multimodal receptor antagonist an antagonist of 5-HT7 5-HT2C 5-HT2A and 5-HT6 receptors (Kis 0.153 1.17 1.56 and 2.04 nM respectively) selective for these receptors over 5-HT2B 5-HT5A 5-HT1A and 5-HT1B receptors (Kis 10.6 20.8 61 and 720 nM respectively) an antagonist of a2A- a2B- a2C- a1B- a1A- and a1D-adrenergic receptors (Kis 0.41 1.77 3.55 9.4 18.9 and 30.2 nM respectively) as well as histamine H1 and H2 receptors (Kis 0.58 and 89 nM respectively) increases the time spent in the open arms of the elevated plus maze in mice at 0.2 1 and 5 mg/kg reduces immobility in the forced swim test and prevents scopolamine-induced amnesia in a passive avoidance test in mice at 0.05 mg/kg
Catalog No. 50-294-2022