Learn More
Cayman Chemical PrednIsolon PhosphatesodIu 25g
SDP
Supplier:  Cayman Chemical 1593325 G
Encompass_Preferred
A prodrug form of prednisolone; decreases inflammation and the levels of IL-1β in the synovium, as well as reduces damage to the cartilage matrix in a mouse model of antigen-induced arthritis when administered at 10 mg/kg in long-circulating liposomes; reduces tumor growth in B16/F10 melanoma and C26 colon carcinoma mouse models at 20 mg/kg in long-circulating liposomes
Catalog No. 50-296-8672