Learn More
Apexbio Technology LLC Prednisolone 50-24-8 10mM (in 1mL DMSO)
SDP
Supplier:  Apexbio Technology LLC B20125.1
Encompass_Preferred
Prednisolone is a synthetic glucocorticoid structurally derived from cortisol characterized by anti-inflammatory and immunosuppressive properties Through binding to intracellular glucocorticoid receptors prednisolone modulates gene expression leading to inhibition of pro-inflammatory cytokine production suppression of immune cell proliferation and attenuation of inflammatory mediators In biomedical research prednisolone is commonly employed to investigate inflammatory pathways autoimmune responses and immune regulation mechanisms Additionally it serves as a reference compound in the screening and evaluation of novel anti-inflammatory and immunomodulatory therapeutics
Catalog No. 50-000-62447