Learn More
Electron Microscopy Sciences Tissue-Prep Buffered 10% Formalin
SDP
Supplier:  Electron Microscopy Sciences 1574220
Encompass_Preferred
Prefilled with 10% formalin Cat# 15740. Ready-to-use in pre-filled cups. Labeled in accordance to OSHA requirements, which gives you immediate information regarding handling and first aid procedures. Pre-measured in wide-mouth, leak-proof, opaque polypropylene containers which limits your exposure to formaldehyde and saves you time. 30ml Cup Size, 20ml filled.
Catalog No. 50-980-506