Learn More
Medchemexpress LLC Pregna-1,4-diene-3,20-dione, 11,17,21-trihydroxy-, (11β)- | 50-24-8 | MFCD00003649 | 99.9% | 360.44 | 500 MG
SDP
Supplier:  Medchemexpress LLC HY17463500MG
Encompass_Preferred
Prednisolone is a potent, orally active corticosteroid and a glucocorticoid. It exhibits about four times the anti-inflammatory activity of hydrocortisone while causing less salt and water retention. It can be used for ocular and anti-inflammatory research, and has been shown to inhibit human leukocyte mitosis.
  • Potent, orally active corticosteroid
  • Glucocorticoid properties
  • Exhibits potent anti-inflammatory activity
  • Causes less salt and water retention
  • Suitable for ocular and anti-inflammatory research
  • Inhibits human leukocyte mitosis
  • High purity (99.93%)
  • White to off-white solid appearance
Catalog No. 50-003-86209