Learn More
Medchemexpress LLC Pregna-1,4-diene-3,20-dione, 21-(acetyloxy)-11-hydroxy-6-methyl-17-(1-oxopropoxy) | 86401-95-8 | MFCD00866150 | 98.9% | 472.57 g/mol | C27H36O7 | 10 MG
SDP
Supplier:  Medchemexpress LLC HY10302510MG
Encompass_Preferred
Methylprednisolone aceponate is a research-grade glucocorticoid analytical standard used as a reference material in analytical and biochemical assays. It is intended for laboratory research use only and is commonly used for method development and quantitation by HPLC, GC, and mass spectrometry. CAS 86401-95-8; molecular formula C27H36O7; molecular weight 472.57 g/mol.
  • Research-grade analytical standard suitable for HPLC, GC, and MS
  • Acts as a selective glucocorticoid receptor ligand for anti-inflammatory studies
  • High reported purity (98.9% by LCMS)
  • Available in small-mass sizes for use as calibration and reference material
  • For research use only; not intended for human or clinical use
Catalog No. 50-002-45954