Learn More
Medchemexpress LLC Pregnanolone sulfate (pyridinium) | 124107-39-7 | 99.8% | 477.66 | C26H39NO5S | 50mg
SDP
Supplier:  Medchemexpress LLC HYW74295750MG
Encompass_Preferred
Pregnanolone sulfate pyridinium is an endogenous neurosteroid that inhibits NMDA receptors and is neuroprotective[1]
Catalog No. 50-002-69292