Learn More
Medchemexpress LLC Pregnenolone | 145-13-1 | 99.9% | 1 G
SDP
Supplier:  Medchemexpress LLC HYB01511G
Encompass_Preferred
Pregnenolone is a powerful neurosteroid and the main precursor of various steroid hormones. It functions as a signaling-specific inhibitor of the cannabinoid CB1 receptor, counteracting the effects of tetrahydrocannabinol (THC) and protecting the brain from cannabis intoxication. It also acts as a TRPM3 channel activator and weakly activates TRPM1 channels.
  • Powerful neurosteroid and precursor of steroid hormones
  • Signaling-specific inhibitor of the cannabinoid CB1 receptor
  • Inhibits effects of tetrahydrocannabinol (THC)
  • Protects the brain from cannabis intoxication
  • TRPM3 channel activator
  • Weakly activates TRPM1 channels
  • For research use only
Catalog No. 50-004-05988