Learn More
Medchemexpress LLC Pregnenolone | 145-13-1 | MFCD00003628 | 99.9% | 10 G
SDP
Supplier:  Medchemexpress LLC HYB015110G
Encompass_Preferred
Pregnenolone is a powerful neurosteroid and the main precursor of various steroid hormones, including steroid ketones. It acts as a signaling-specific inhibitor of the cannabinoid CB1 receptor and can protect the brain from cannabis intoxication. Additionally, it functions as a TRPM3 channel activator and can weakly activate TRPM1 channels.
  • Powerful neurosteroid
  • Main precursor of various steroid hormones
  • Signaling-specific inhibitor of the cannabinoid CB1 receptor
  • Protects the brain from cannabis intoxication
  • TRPM3 channel activator
  • Weakly activates TRPM1 channels
Catalog No. 50-004-07234