Learn More
Medchemexpress LLC Pregnenolone (3β-Hydroxy-5-pregnen-20-one) | 145-13-1 | 99.9% | 1 ML
SDP
Supplier:  Medchemexpress LLC HYB015110MM/1ML
Encompass_Preferred
Pregnenolone is a potent neurosteroid and the primary precursor to various steroid hormones, including steroid ketones. It functions as a signaling-specific inhibitor of the cannabinoid CB1 receptor, thereby inhibiting the effects of tetrahydrocannabinol (THC) that are mediated by CB1 receptors. Pregnenolone has the ability to protect the brain from cannabis intoxication. Additionally, it acts as a TRPM3 channel activator and can weakly activate TRPM1 channels.
  • Powerful neurosteroid
  • Main precursor of various steroid hormones
  • Signaling-specific inhibitor of cannabinoid CB1 receptor
  • Inhibits THC effects mediated by CB1 receptors
  • Can protect the brain from cannabis intoxication
  • TRPM3 channel activator
  • Weakly activates TRPM1 channels
Catalog No. 50-004-06303