Learn More
Medchemexpress LLC Pregnenolone acetate | 1778-02-5 | MFCD00003627 | 98.4% | 100 G
SDP
Supplier:  Medchemexpress LLC HYB1740100G
Encompass_Preferred
Pregnenolone acetate is a nonmetabolized analog of Pregnenolone that promotes neurite extension and induces changes in growth cone morphology in primary cultures of cerebellar granule neurons. As a neurosteroid, it increases microtubule polymerization and holds potential for neurodevelopmental diseases research.
  • Promotes neurite extension
  • Induces change in growth cone morphology
  • Increases microtubule polymerization
  • Potential for neurodevelopmental diseases research
  • Used in hormone replacement therapy research
  • Explored for neuroprotection in diseases like Alzheimer's
  • Suggested to modulate stress responses
  • Investigated for anti-inflammatory applications
  • Utilized in cosmetic formulations for skin health
Catalog No. 50-004-13185