Learn More
Electron Microscopy Sciences Premiere Scalpel Blade #11 | 100/PK
SDP
Supplier:  Electron Microscopy Sciences 7204911
Encompass_Preferred
Premiere Blades are made from carbon steel, individually foil wrapped and gamma radiation sterilized. They are exceptionally strong and relatively uniformity sharpwhich is ideal for routine laboratory dissecting, tissue mincing and where you do not need high cost blades.
Catalog No. 50-190-0379