Learn More
Yamazen Science Inc PREMIUM UNIVERSAL COLUMN M
Supplier:  Yamazen Science Inc UW10201
Encompass
This item has a minimum qty of 3 per supplier requirements.
Premium Universal Column: High Throughput Purification PerformancePore Size 60 angstroms, Particle Size 25/40um, Silica Gel, Luer Lock TypeSize: M10/pk
Catalog No. NC1465387