Learn More
Takara Bio Premix Taq™ DNA Polymerase (TaKaRa Taq™ Version 2.0), 150 units
SDP
Supplier:  Takara Bio R004A
Encompass
A convenient 2X PCR master mix of Takara Taq enzyme, buffer, and dNTP mixture, which simplifies the setup of PCR reactions and minimizes pipetting steps. 150 units
Catalog No. 50-443-954