Learn More
Waters Corp Nova-Pak C18 Prep Column, 60Å, 6 µm, 19 mm X 300 mm, 1/pk
Supplier:  Waters Corp WAT025822
Encompass_Preferred
Nova-Pak HR C18 columns are silica-based, semi-preparative and preparative columns that are based on 6 µm particle technology. Compared to a 5 µm column, Nova-Pak HR columns offer higher resolution, more efficient separations, and faster chromatography.
Catalog No. 50-785-212