Learn More
Waters Corp Nova-Pak Guard, C18, 6 μm, 60 Å, 25 mm x 10 mm
Supplier: Waters Corp WAT038528
Nova-Pak HR C18 columns are silica-based, semi-preparative and preparative columns that utilize 6 μm particle technology. These columns offer higher resolution, more efficient separations, and faster chromatography.
- C18 chemistry for reversed phase separation.
- Silica particle substrate with spherical particle shape.
- Particle size of 6 μm and pore size of 60 Å.
- pH range of 2 to 8 pH.
- Maximum pressure of 6000 psi (415 Bar).
- Endcapped for improved performance.
- Medium silanol activity.
- Radial-Pak guard format.
- Designed for HPLC systems.
- Suitable for peptide applications.
- Inner diameter of 25 mm and length of 10 mm.
- Units per package: 2 pk.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.