Learn More
Pion Inc Pion PrepTube 250 mL, Clear Polypropylene Tube, Stainless Steel Blade, 10 per Pack
Supplier:  Pion Inc SP05000501
Encompass
The Pion Inc. PrepTube 250 mL is a clear polypropylene tube with a stainless steel blade, enhancing efficiency in sample prep for scientific applications. It accelerates preparation time by up to 90%, providing robust mixing and compatibility with diverse lab workflows. Designed for single-use, each tube includes a cap with a hole for easy access, making it ideal for tasks like assay and impurity analyses.
Catalog No. NC3359659