Learn More
LAMDA BIOTECH INC PreSafeStain 6X DNA Load Dye
Supplier:  LAMDA BIOTECH INC 6XLD
Encompass_Preferred
A multi-functional DNA loading buffer containing integrated fluorescent dye for direct DNA visualization during gel electrophoresis. Features dual-color tracking system and green fluorescent DNA stain in a single solution.
Catalog No. 50-204-5814