Learn More
AURORA BIOLABS LLC PreScission Protease (HRV 3C), 1000 units
Supplier:  AURORA BIOLABS LLC 1900021K
Encompass
PreScission Protease is a fusion protein of glutathione S-transferase (GST) and human rhinovirus (HRV) type 14 3C protease. The protease specifically recognizes a subset of sequences which include the core amino acid sequence Leu-Phe-Gln/Gly-Pro cleaving between the Gln and Gly residues. Substrate recognition and cleavage are likely to be dependent not only upon primary structural signals, but also upon the secondary and tertiary structures of the fusion protein as well.
Catalog No. NC3787970