Learn More
Pendotech Pendotech Pressure Sensor Transmitter, 30 Psi
Supplier:  Pendotech PT30
Encompass
Pressure sensor transmitter with 4-20mA output, 30 psi (2 bar) DIN rail mount, 24VDC, with calibration certificate and reusable sensor cable installed
Catalog No. NC1831945
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.